Assay Details
| Gene Symbol: h2az2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153644.1 | NP_705930.1 | 4 - 5 | 373 | 105 | |
| GenBank mRNA | AF414110.1 | - | 4 - 5 | 373 | 105 | |
| BC049019.1 | - | 4 - 5 | 401 | 105 | ||
| BC154791.1 | - | 4 - 5 | 401 | 105 | ||
| EH508860.1 | - | 4 - 5 | 468 | 105 | ||
More Information
Set Membership: |
|

h2az2a
Genomic Map