Assay Details
| Gene Symbol: il17rd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328553.2 | NP_001315482.2 | 12 - 13 | 2197 | 92 | |
| NM_153660.3 | NP_705946.3 | 12 - 13 | 2200 | 92 | ||
| GenBank mRNA | AF364103.1 | - | 12 - 13 | 2142 | 92 | |
| AF401232.1 | - | 12 - 13 | 2131 | 92 | ||
| BC128813.1 | - | 12 - 13 | 2122 | 92 | ||
| BC163933.1 | - | 12 - 13 | 2203 | 92 | ||
More Information
Set Membership: |
|

il17rd
Genomic Map