Assay Details
| Gene Symbol: tbx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153665.1 | NP_705951.1 | 1 - 2 | 444 | 71 | |
| GenBank mRNA | AF448503.3 | - | 1 - 2 | 432 | 71 | |
| BC154826.1 | - | 1 - 2 | 428 | 71 | ||
| BC163460.1 | - | 1 - 2 | 389 | 71 | ||
| BC163482.1 | - | 1 - 2 | 389 | 71 | ||
More Information
Set Membership: |
|

tbx18
Genomic Map