Assay Details
| Gene Symbol: tbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153666.2 | NP_705952.2 | 3 - 4 | 662 | 68 | |
| GenBank mRNA | AB036516.1 | - | 3 - 4 | 662 | 68 | |
| BC163297.1 | - | 3 - 4 | 550 | 68 | ||
More Information
Set Membership: |
|

tbx6
Genomic Map