Assay Details
| Gene Symbol: adssl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173237.2 | NP_775344.2 | 12 - 13 | 1474 | 101 | |
| GenBank mRNA | AY099534.1 | - | 12 - 13 | 1348 | 101 | |
| BC051613.1 | - | 12 - 13 | 1334 | 101 | ||
More Information
Set Membership: |
|

adssl
Genomic Map