Assay Details
| Gene Symbol: adssl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173237.2 | NP_775344.2 | 4 - 5 | 571 | 80 | |
| GenBank mRNA | AY099534.1 | - | 4 - 5 | 445 | 80 | |
| BC051613.1 | - | 4 - 5 | 433 | 80 | ||
| Gene Symbol: zgc:111868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC055595.1 | - | 461 | 80 | ||
More Information
Set Membership: |
|

adssl
Genomic Map