Assay Details
| Gene Symbol: med29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173271.1 | NP_775378.1 | 2 - 3 | 243 | 86 | |
| GenBank mRNA | AF506210.1 | - | 2 - 3 | 243 | 86 | |
| BC094992.1 | - | 2 - 3 | 239 | 86 | ||
| BC164953.1 | - | 2 - 3 | 264 | 86 | ||
More Information
Set Membership: |
|

med29
Genomic Map