Assay Details
| Gene Symbol: plp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175576.2 | NP_783166.1 | 5 - 6 | 627 | 80 | |
| GenBank mRNA | AY070259.1 | - | 5 - 6 | 627 | 80 | |
| BC095297.1 | - | 5 - 6 | 629 | 80 | ||
| BC165098.1 | - | 4 - 5 | 620 | 80 | ||
More Information
Set Membership: |
|

plp1a
Genomic Map