Assay Details
| Gene Symbol: selenop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178297.4 | NP_840082.3 | 3 - 4 | 508 | 71 | |
| GenBank mRNA | AF322071.1 | - | 3 - 4 | 492 | 71 | |
| BC059656.1 | - | 2 - 3 | 71 | |||
| BC063960.1 | - | 2 - 3 | 448 | 71 | ||
| BC155821.1 | - | 3 - 4 | 481 | 71 | ||
| BC159239.1 | - | 3 - 4 | 381 | 71 | ||
| BC159240.1 | - | 3 - 4 | 481 | 71 | ||
| CA473236.1 | - | 3 - 4 | 71 | |||
| CD759036.1 | - | 3 - 4 | 669 | 71 | ||
More Information
Set Membership: |
|

selenop
Genomic Map