Assay Details
| Gene Symbol: selenop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001353911.1 | NP_001340840.1 | 4 - 5 | 651 | 87 | |
| GenBank mRNA | AF322072.1 | - | 4 - 5 | 593 | 87 | |
| BG985559.1 | - | 2 - 3 | 223 | 87 | ||
| EH498018.1 | - | 4 - 5 | 683 | 87 | ||
| EH518168.1 | - | 1 - 2 | 676 | 87 | ||
| EH527207.1 | - | 1 - 2 | 675 | 87 | ||
More Information
Set Membership: |
|

selenop2
Genomic Map