Assay Details
| Gene Symbol: anxa1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181760.2 | NP_861425.1 | 5 - 6 | 487 | 97 | |
| XM_005165606.4 | XP_005165663.1 | 6 - 7 | 560 | 97 | ||
| GenBank mRNA | AY178795.1 | - | 5 - 6 | 492 | 97 | |
More Information
Set Membership: |
|

anxa1c
Genomic Map