Assay Details
| Gene Symbol: anxa11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181766.2 | NP_861431.1 | 4 - 5 | 607 | 61 | |
| GenBank mRNA | AY178802.2 | - | 4 - 5 | 607 | 61 | |
| BC068366.1 | - | 4 - 5 | 590 | 61 | ||
More Information
Set Membership: |
|

anxa11b
Genomic Map