Assay Details
| Gene Symbol: anxa11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181766.2 | NP_861431.1 | 5 - 6 | 705 | 68 | |
| GenBank mRNA | AY178802.2 | - | 5 - 6 | 705 | 68 | |
| BC068366.1 | - | 5 - 6 | 688 | 68 | ||
More Information
Set Membership: |
|

anxa11b
Genomic Map