Assay Details
| Gene Symbol: fgf10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182870.2 | NP_878290.1 | 1 - 2 | 975 | 70 | |
| GenBank mRNA | AB091000.1 | - | 1 - 2 | 296 | 70 | |
| AF544025.1 | - | 1 - 2 | 979 | 70 | ||
| BC094986.1 | - | 1 - 2 | 949 | 70 | ||
| BC165003.1 | - | 1 - 2 | 339 | 70 | ||
More Information
Set Membership: |
|

fgf10a
Genomic Map