Assay Details
| Gene Symbol: egr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328404.2 | NP_001315333.1 | 2 | 797 | 72 | |
| NM_001359896.1 | NP_001346825.1 | 2 | 612 | 72 | ||
| GenBank mRNA | AY070229.1 | - | 1 | 1053 | 72 | |
| BC058293.1 | - | 2 | 557 | 72 | ||
| BC163879.1 | - | 1 | 328 | 72 | ||
| BC163884.1 | - | 1 | 328 | 72 | ||
More Information
Set Membership: |
|

egr2a
Genomic Map