Assay Details
| Gene Symbol: hnf1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194372.3 | NP_919353.1 | 8 - 9 | 1614 | 72 | |
| GenBank mRNA | AF250352.1 | - | 8 - 9 | 1614 | 72 | |
More Information
Set Membership: |
|

hnf1bb
Genomic Map