Assay Details
| Gene Symbol: arpc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194378.4 | NP_919359.2 | 2 - 3 | 309 | 68 | |
| GenBank mRNA | AF139537.1 | - | 2 - 3 | 295 | 68 | |
| BC044413.1 | - | 255 | 68 | |||
| BC070012.1 | - | 2 - 3 | 234 | 68 | ||
| BC164286.1 | - | 2 - 3 | 253 | 68 | ||
| EH437356.1 | - | 2 - 3 | 338 | 68 | ||
More Information
Set Membership: |
|

arpc5a
Genomic Map