Assay Details
| Gene Symbol: mmp14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194414.1 | NP_919395.1 | 8 - 9 | 1657 | 66 | |
| XM_005171310.4 | XP_005171367.1 | 9 - 10 | 1585 | 66 | ||
| GenBank mRNA | AY324197.1 | - | 8 - 9 | 1657 | 66 | |
| BC162135.1 | - | 8 - 9 | 1549 | 66 | ||
More Information
Set Membership: |
|

mmp14b
Genomic Map