Assay Details
| Gene Symbol: slc15a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198064.1 | NP_932330.1 | 22 - 23 | 2045 | 61 | |
| GenBank mRNA | AY300011.1 | - | 22 - 23 | 2045 | 61 | |
| BC054899.1 | - | 22 - 23 | 2045 | 61 | ||
More Information
Set Membership: |
|

slc15a1b
Genomic Map