Assay Details
| Gene Symbol: slc15a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198064.1 | NP_932330.1 | 8 - 9 | 769 | 119 | |
| GenBank mRNA | AY300011.1 | - | 8 - 9 | 769 | 119 | |
| BC054899.1 | - | 8 - 9 | 769 | 119 | ||
More Information
Set Membership: |
|

slc15a1b
Genomic Map