Assay Details
| Gene Symbol: tpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198140.1 | NP_937783.1 | 4 - 5 | 469 | 171 | |
| GenBank mRNA | AF288217.1 | - | 4 - 5 | 451 | 171 | |
| BC049059.1 | - | 4 - 5 | 469 | 171 | ||
More Information
Set Membership: |
|

tpt1
Genomic Map