Assay Details
| Gene Symbol: taf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198368.1 | NP_938182.1 | 2 - 3 | 322 | 69 | |
| XM_009292430.3 | XP_009290705.1 | 2 - 3 | 315 | 69 | ||
| GenBank mRNA | BC050160.1 | - | 2 - 3 | 348 | 69 | |
| BC056696.1 | - | 2 - 3 | 322 | 69 | ||
| BC065878.1 | - | 2 - 3 | 302 | 69 | ||
| BC152198.1 | - | 2 - 3 | 352 | 69 | ||
More Information
Set Membership: |
|

taf12
Genomic Map