Assay Details
| Gene Symbol: stap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198808.1 | NP_942103.1 | 1 - 2 | 127 | 118 | |
| GenBank mRNA | BC054569.1 | - | 1 - 2 | 127 | 118 | |
More Information
Set Membership: |
|

stap2a
Genomic Map