Assay Details
| Gene Symbol: stap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198808.1 | NP_942103.1 | 3 - 4 | 322 | 93 | |
| XM_021478992.1 | XP_021334667.1 | 2 - 3 | 629 | 93 | ||
| GenBank mRNA | BC054569.1 | - | 3 - 4 | 322 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021478992.1 transcripts.
Set Membership: |
|

stap2a
Genomic Map