Assay Details
| Gene Symbol: scd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198815.2 | NP_942110.2 | 3 - 4 | 568 | 93 | |
| GenBank mRNA | AY217090.1 | - | 2 - 3 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scd| Gene Symbol: scd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198815.2 | NP_942110.2 | 3 - 4 | 568 | 93 | |
| GenBank mRNA | AY217090.1 | - | 2 - 3 | 93 | ||
Set Membership: |
|