Assay Details
| Gene Symbol: p2rx4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021478206.1 | XP_021333881.1 | 1 - 2 | 601 | 81 | |
| GenBank mRNA | AY292652.1 | - | 1 - 2 | 135 | 81 | |
More Information
Set Membership: |
|

p2rx4b
Genomic Map