Assay Details
| Gene Symbol: p2rx4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198987.2 | NP_945338.2 | 7 - 8 | 775 | 114 | |
| XM_021478206.1 | XP_021333881.1 | 7 - 8 | 1227 | 114 | ||
| GenBank mRNA | AY292652.1 | - | 7 - 8 | 770 | 114 | |
| BC162774.1 | - | 7 - 8 | 775 | 114 | ||
More Information
Set Membership: |
|

p2rx4b
Genomic Map