Assay Details
| Gene Symbol: atad5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082829.2 | NP_001076298.2 | 8 - 9 | 1993 | 71 | |
| XM_005166177.4 | XP_005166234.1 | 8 - 9 | 2049 | 71 | ||
| GenBank mRNA | BC057535.1 | - | 8 - 9 | 1993 | 71 | |
More Information
Set Membership: |
|

atad5b
Genomic Map