Assay Details
| Gene Symbol: mier1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199559.1 | NP_955853.1 | 10 - 11 | 973 | 87 | |
| XM_021476778.1 | XP_021332453.1 | 12 - 13 | 1224 | 87 | ||
| XM_021476779.1 | XP_021332454.1 | 12 - 13 | 1221 | 87 | ||
| GenBank mRNA | BC045973.1 | - | 10 - 11 | 973 | 87 | |
| BC164544.1 | - | 9 - 10 | 933 | 87 | ||
More Information
Set Membership: |
|

mier1a
Genomic Map