Assay Details
| Gene Symbol: mier1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199559.1 | NP_955853.1 | 12 - 13 | 1240 | 97 | |
| XM_017356435.2 | XP_017211924.1 | 14 - 15 | 1406 | 97 | ||
| XM_017356436.2 | XP_017211925.1 | 14 - 15 | 1400 | 97 | ||
| XM_021476778.1 | XP_021332453.1 | 14 - 15 | 1491 | 97 | ||
| XM_021476779.1 | XP_021332454.1 | 14 - 15 | 1488 | 97 | ||
| GenBank mRNA | BC045973.1 | - | 12 - 13 | 1240 | 97 | |
| BC164544.1 | - | 11 - 12 | 1200 | 97 | ||
More Information
Set Membership: |
|

mier1a
Genomic Map