Assay Details
| Gene Symbol: mier1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199559.1 | NP_955853.1 | 2 - 3 | 112 | 116 | |
| XM_017356435.2 | XP_017211924.1 | 4 - 5 | 359 | 116 | ||
| XM_017356436.2 | XP_017211925.1 | 4 - 5 | 356 | 116 | ||
| XM_021476778.1 | XP_021332453.1 | 4 - 5 | 363 | 116 | ||
| XM_021476779.1 | XP_021332454.1 | 4 - 5 | 360 | 116 | ||
| GenBank mRNA | BC045973.1 | - | 2 - 3 | 112 | 116 | |
| BC164544.1 | - | 1 - 2 | 72 | 116 | ||
More Information
Set Membership: |
|

mier1a
Genomic Map