Assay Details
| Gene Symbol: cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199598.1 | NP_955892.1 | 1 - 2 | 153 | 73 | |
| GenBank mRNA | BC045362.1 | - | 1 - 2 | 153 | 73 | |
| BC165743.1 | - | 1 - 2 | 83 | 73 | ||
| BC166040.1 | - | 1 - 2 | 149 | 73 | ||
More Information
Set Membership: |
|

cs
Genomic Map