Assay Details
| Gene Symbol: dnajc5ga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199623.1 | NP_955917.1 | 4 - 5 | 669 | 64 | |
| XM_005160813.4 | XP_005160870.1 | 4 - 5 | 912 | 64 | ||
| GenBank mRNA | BC049534.1 | - | 4 - 5 | 669 | 64 | |
More Information
Set Membership: |
|

dnajc5ga
Genomic Map