Assay Details
| Gene Symbol: znf207b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199665.2 | NP_955959.1 | 1 - 2 | 161 | 97 | |
| XM_009302342.3 | XP_009300617.1 | 1 - 2 | 172 | 97 | ||
| XM_021476738.1 | XP_021332413.1 | 1 - 2 | 173 | 97 | ||
| GenBank mRNA | AY422996.1 | - | 1 - 2 | 106 | 97 | |
| BC049335.1 | - | 1 - 2 | 164 | 97 | ||
| BC065453.1 | - | 1 - 2 | 127 | 97 | ||
More Information
Set Membership: |
|

znf207b
Genomic Map