Assay Details
| Gene Symbol: znf207b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199665.2 | NP_955959.1 | 9 - 10 | 1275 | 107 | |
| XM_009302342.3 | XP_009300617.1 | 10 - 11 | 1379 | 107 | ||
| XM_021476738.1 | XP_021332413.1 | 10 - 11 | 1380 | 107 | ||
| GenBank mRNA | AY422996.1 | - | 9 - 10 | 1220 | 107 | |
| BC049335.1 | - | 9 - 10 | 1278 | 107 | ||
| BC065453.1 | - | 9 - 10 | 1241 | 107 | ||
More Information
Set Membership: |
|

znf207b
Genomic Map