Assay Details
| Gene Symbol: hmox1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127516.1 | NP_001120988.1 | 3 - 4 | 396 | 74 | |
| GenBank mRNA | BC061954.1 | - | 3 - 4 | 216 | 74 | |
More Information
Set Membership: |
|

hmox1a
Genomic Map