Assay Details
| Gene Symbol: cpne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276281.1 | NP_001263210.1 | 15 - 16 | 1618 | 61 | |
| NM_199699.2 | NP_955993.2 | 15 - 16 | 1615 | 61 | ||
| XM_005169257.4 | XP_005169314.1 | 15 - 16 | 1574 | 61 | ||
| XM_005169258.4 | XP_005169315.1 | 15 - 16 | 1570 | 61 | ||
| XM_005169260.4 | XP_005169317.1 | 15 - 16 | 1607 | 61 | ||
| XM_005169262.3 | XP_005169319.1 | 15 - 16 | 1604 | 61 | ||
| XM_009306020.2 | XP_009304295.1 | 15 - 16 | 1570 | 61 | ||
| XM_017358205.2 | XP_017213694.1 | 15 - 16 | 1509 | 61 | ||
| GenBank mRNA | AY423010.1 | - | 15 - 16 | 1550 | 61 | |
| BC050240.1 | - | 15 - 16 | 1608 | 61 | ||
| BC151889.1 | - | 15 - 16 | 1577 | 61 | ||
| BC164658.1 | - | 14 - 15 | 1518 | 61 | ||
More Information
Set Membership: |
|

cpne1
Genomic Map