Assay Details
| Gene Symbol: pigp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199712.2 | NP_956006.2 | 2 - 3 | 146 | 96 | |
| XM_005172570.4 | XP_005172627.1 | 2 - 3 | 599 | 96 | ||
| GenBank mRNA | BC059676.1 | - | 1 - 2 | 111 | 96 | |
More Information
Set Membership: |
|

pigp
Genomic Map