Assay Details
| Gene Symbol: bin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199796.1 | NP_956090.1 | 11 - 12 | 1547 | 104 | |
| XM_005166011.4 | XP_005166068.1 | 10 - 11 | 1669 | 104 | ||
| GenBank mRNA | BC044473.1 | - | 11 - 12 | 1547 | 104 | |
More Information
Set Membership: |
|

bin2b
Genomic Map