Assay Details
| Gene Symbol: gnsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199841.2 | NP_956135.2 | 10 - 11 | 1229 | 104 | |
| XM_021479111.1 | XP_021334786.1 | 10 - 11 | 1126 | 104 | ||
| GenBank mRNA | BC045323.1 | - | 10 - 11 | 1229 | 104 | |
More Information
Set Membership: |
|

gnsb
Genomic Map