Assay Details
| Gene Symbol: wrap73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328537.1 | NP_001315466.1 | 9 - 10 | 995 | 62 | |
| GenBank mRNA | BC050515.1 | - | 9 - 10 | 996 | 62 | |
| BC154787.1 | - | 9 - 10 | 942 | 62 | ||
More Information
Set Membership: |
|

wrap73
Genomic Map