Assay Details
| Gene Symbol: ankmy2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199894.1 | NP_956188.1 | 7 - 8 | 830 | 87 | |
| GenBank mRNA | BC044410.1 | - | 7 - 8 | 830 | 87 | |
More Information
Set Membership: |
|

ankmy2a
Genomic Map