Assay Details
| Gene Symbol: aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199910.2 | NP_956204.2 | 4 - 5 | 523 | 82 | |
| XM_005161011.4 | XP_005161068.1 | 4 - 5 | 603 | 82 | ||
| XM_021468757.1 | XP_021324432.1 | 4 - 5 | 735 | 82 | ||
| GenBank mRNA | BC056802.1 | - | 4 - 5 | 599 | 82 | |
| FJ655385.1 | - | 4 - 5 | 523 | 82 | ||
More Information
Set Membership: |
|

aqp7
Genomic Map