Assay Details
| Gene Symbol: polr2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167956.2 | NP_001161428.1 | 2 - 3 | 323 | 68 | |
| GenBank mRNA | BC046047.1 | - | 2 - 3 | 235 | 68 | |
| EH524379.1 | - | 7 - 8 | 793 | 68 | ||
| EH555113.1 | - | 2 - 3 | 367 | 68 | ||
More Information
Set Membership: |
|

polr2c
Genomic Map