Assay Details
| Gene Symbol: tada3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199926.1 | NP_956220.1 | 3 - 4 | 524 | 97 | |
| GenBank mRNA | BC055157.1 | - | 3 - 4 | 524 | 97 | |
More Information
Set Membership: |
|

tada3l
Genomic Map