Assay Details
| Gene Symbol: uck2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171067.1 | NP_001164538.1 | 3 - 4 | 398 | 85 | |
| GenBank mRNA | BC054665.1 | - | 3 - 4 | 398 | 85 | |
| CK694919.1 | - | 1 - 2 | 109 | 85 | ||
| CK695146.1 | - | 1 - 2 | 182 | 85 | ||
More Information
Set Membership: |
|

uck2a
Genomic Map