Assay Details
| Gene Symbol: uck2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171067.1 | NP_001164538.1 | 6 - 7 | 688 | 139 | |
| GenBank mRNA | BC054665.1 | - | 6 - 7 | 688 | 139 | |
| CK694919.1 | - | 4 - 5 | 399 | 140 | ||
More Information
Set Membership: |
|

uck2a
Genomic Map