Assay Details
| Gene Symbol: cldnd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199961.1 | NP_956255.1 | 1 - 2 | 536 | 57 | |
| GenBank mRNA | BC060671.1 | - | 1 - 2 | 536 | 57 | |
More Information
Set Membership: |
|

cldnd1a
Genomic Map