Assay Details
| Gene Symbol: rbp4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199965.1 | NP_956259.1 | 4 - 5 | 434 | 113 | |
| GenBank mRNA | BC059516.1 | - | 4 - 5 | 434 | 113 | |
| BC152209.1 | - | 4 - 5 | 457 | 113 | ||
| BC152210.1 | - | 4 - 5 | 508 | 113 | ||
| KM655654.1 | - | 3 - 4 | 371 | 113 | ||
More Information
Set Membership: |
|

rbp4l
Genomic Map