Assay Details
| Gene Symbol: rbp4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199965.1 | NP_956259.1 | 5 - 6 | 640 | 85 | |
| GenBank mRNA | BC059516.1 | - | 5 - 6 | 640 | 85 | |
| BC152209.1 | - | 5 - 6 | 663 | 85 | ||
| BC152210.1 | - | 5 - 6 | 714 | 85 | ||
More Information
Set Membership: |
|

rbp4l
Genomic Map